Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Vinculin, Mouse anti-Human,Mouse, Clone: VCL/2573, Novus Biologicals™

Mouse Monoclonal Antibody
$280.00 - $536.00
Specifications
Antigen | Vinculin |
---|---|
Clone | VCL/2573 |
Concentration | 0.2 mg/mL |
Dilution | Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array |
Description
Vinculin Monoclonal specifically detects Vinculin in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.Specifications
Vinculin | |
0.2 mg/mL | |
Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array | |
Unconjugated | |
Mouse | |
Cell Biology, Cellular Markers, Phospho Specific | |
10 mM PBS with 0.05% BSA | |
CMD1W, CMH15, Metavinculin, MVCL, vinculin | |
VCL | |
IgG1 κ | |
Protein A or G purified |
VCL/2573 | |
Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse | |
P18206, P18206 | |
7414 | |
Recombinant fragment (around aa 174-322) of human Vinculin protein (exact sequence is proprietary) | |
Primary | |
Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title