Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vinculin Mouse anti-Human, Mouse, Porcine, Rat, Clone: 2B5A7, Proteintech
SDP

Mouse Monoclonal Antibody

Supplier:  Proteintech Group Inc 663051IG150UL

Encompass_Preferred

Catalog No. 50-106-0619


May include imposed supplier surcharges.
Only null left
Add to Cart

Description

Description

Vinculin (VCL) is a cytoskeletal protein associated with cell-cell and cell-extracellular matrix adherens-type junctions. It functions as one of several interacting proteins involved in anchoring F-actin to the membrane. It has been shown that a sequence of molecular interactions might be involved in the transmembrane assembly of adhesion plaques. In the assembly of adhesion plaques, the beta subunit of integrin binds to talin. Talin binds to vinculin that interacts with alpha-actinin and possibly with itself. Since alpha-actinin binds to and cross-links actin filaments, vinculin represents a key element in the transmembrane linkage of the extracellular matrix to the cytoplasmic microfilament system. Mutations in the gene can result in cardiomyopathy dilated 1W or familial hypertrophic 15.
Specifications

Specifications

Vinculin
Monoclonal
0.78 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P18206, P26234, P85972, Q64727
VCL
Vinculin Fusion Protein Ag24946
150 μL
Primary
Human, Mouse, Pig, Rat
Antibody
IgG1
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
2B5A7
Unconjugated
VCL
CMD1W, Metavinculin, MVCL, VCL, Vinculin
Mouse
Protein G
RUO
22330, 305679, 396974, 7414
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.