Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Vinculin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582003
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582003 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582003 Supplier Invitrogen™ Supplier No. PA582003
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KMIDERQQEL THQEHRVMLV NSMNTVKELL PVLISAMKIF VTTKNSKNQG IEEALKNRNF TVEKMSAEIN EIIRVLQLTS WDEDAWASKD TEAMKRALAS IDSKLNQAKG WLRDPSASPG DAGEQAIRQI LDEAGKVGEL CAGKERREI Highest antigen sequence indentity to the following orthologs: Rat - 99%, Mouse - 99%.

Vinculin (VCL) is a cytoskeletal protein associated with cell-cell and cell-extracellular matrix adherens-type junctions. It functions as one of several interacting proteins involved in anchoring F-actin to the membrane. It has been shown that a sequence of molecular interactions might be involved in the transmembrane assembly of adhesion plaques. In the assembly of adhesion plaques, the beta subunit of integrin binds to talin. Talin binds to vinculin that interacts with alpha-actinin and possibly with itself. Since alpha-actinin binds to and cross-links actin filaments, vinculin represents a key element in the transmembrane linkage of the extracellular matrix to the cytoplasmic microfilament system. Mutations in the gene can result in cardiomyopathy dilated 1W or familial hypertrophic 15.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Vinculin
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene VCL
Gene Accession No. P18206, P85972, Q64727
Gene Alias 9430097D22; AA571387; AI462105; AW545629; CMD1W; CMH15; epididymis luminal protein 114; HEL114; metavinculin; meta-vinculin; metavinculin {ECO:0000250; MV; MVCL; RP11-178G16.3; UniProtKB:P18206}; unnamed protein product; Vcl; vcl {ECO:0000250; VIN; vinc; vinc 1; VINC1; Vinculin; vinculin {ECO:0000250
Gene Symbols VCL
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human VCL. Recombinant protein control fragment (Product #RP-100519).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22330, 305679, 7414
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.