Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Vinculin Rabbit anti-Human, Clone: 5G7, Bioss

Rabbit Monoclonal Antibody
Supplier: Bioss BSM54148R
Description
Vinculin (VCL) is a cytoskeletal protein associated with cell-cell and cell-extracellular matrix adherens-type junctions. It functions as one of several interacting proteins involved in anchoring F-actin to the membrane. It has been shown that a sequence of molecular interactions might be involved in the transmembrane assembly of adhesion plaques. In the assembly of adhesion plaques, the beta subunit of integrin binds to talin. Talin binds to vinculin that interacts with alpha-actinin and possibly with itself. Since alpha-actinin binds to and cross-links actin filaments, vinculin represents a key element in the transmembrane linkage of the extracellular matrix to the cytoplasmic microfilament system. Mutations in the gene can result in cardiomyopathy dilated 1W or familial hypertrophic 15.Specifications
| Vinculin | |
| Monoclonal | |
| 1 μg/mL | |
| TBS with 1% BSA, 40% glycerol and 0.05% sodium azide; pH | |
| P18206 | |
| VCL | |
| Synthetic peptide within human Vinculin aa 1000-1100. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunofluorescence, Western Blot | |
| 5G7 | |
| Unconjugated | |
| VCL | |
| 9430097D22; AA571387; AI462105; AW545629; CMD1W; CMH15; epididymis luminal protein 114; HEL114; Metavinculin; metavinculin {ECO:0000250; MV; MVCL; RP11-178G16.3; UniProtKB:P18206}; VCL; vcl {ECO:0000250; VINC1; Vinculin; vinculin {ECO:0000250 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 7414 | |
| -20°C | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction