Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Vinculin Rabbit anti-Human, Mouse, Rat, Clone: JM42-43, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP275708
Description
Vinculin Monoclonal specifically detects Vinculin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.Specifications
Vinculin | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:10 - 1:500 | |
CMD1W, CMH15, Metavinculin, MVCL, vinculin | |
Rabbit | |
124 kDa | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
JM42-43 | |
Unconjugated | |
TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide | |
VCL | |
Synthetic peptide within human Vinculin aa 1000-1060. (SwissProt: P18206 Human; SwissProt: Q64727 Mouse; SwissProt: P85972 Rat) | |
Protein A purified | |
Cellular Markers, Cytoskeleton Markers | |
7414.0 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction