Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Vinexin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111265
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111265 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111265 Supplier Invitrogen™ Supplier No. PA5111265
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YRRMFQQIHR KMPDLQLDWT FEEPPRDPRH LGAQQRPAHR PGPATSSSGR SWDHSEELPR STFNYRPGAF STVLQPSNQV LRRREKVDNV WTEESWNQFL QELETGQRPK KPLVDDPGEK PSQPIE Highest antigen sequence indentity to the following orthologs: Mouse - 68%, Rat - 68%.

The SORBS3 gene encodes 2 isoforms vinexin-alpha and -beta that are vinculin (VCL; MIM 193065)-binding cytoskeletal proteins involved in focal adhesion and cell-cell adhesion (Kioka et al., 1999 [PubMed 9885244]).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Vinexin
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SORBS3
Gene Accession No. O60504
Gene Alias Scam1; SCAM-1; SH3 domain protein 4; SH3 domain-containing protein SH3P3; SH3-containing adapter molecule 1; Sh3d4; SH3P3; sorbin and SH3 domain containing 3; sorbin and SH3 domain-containing protein 3; Sorbs3; Vinexin; vinexin alpha; vinexin beta; vinexin beta (SH3-containing adaptor molecule-1); vinexin-g
Gene Symbols SORBS3
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human Vinexin. Recombinant protein control fragment (Product #RP-106165).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10174
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.