Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vinyl 2-Ethylhexanoate (stabilized with MEHQ) 98.0+%, TCI America™
SDP

Catalog No. E040425ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E040425ML 25 mL
E0404500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. E040425ML Supplier TCI America Supplier No. E040425ML
Add to Cart
Add to Cart
This item is not returnable. View return policy

Chemical Identifiers

CAS 94-04-2
Molecular Formula C10H18O2
Molecular Weight (g/mol) 170.252
MDL Number MFCD00009488
InChI Key IGBZOHMCHDADGY-UHFFFAOYSA-N
Synonym 2-Ethylhexanoic Acid Vinyl Ester
PubChem CID 62343
IUPAC Name ethenyl 2-ethylhexanoate
SMILES CCCCC(CC)C(=O)OC=C

Specifications

Color Colorless
Boiling Point 86°C
Quantity 25 mL
Formula Weight 170.25
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Vinyl 2-Ethylhexanoate (stabilized with MEHQ)
EINECSNumber (2)-0761
RTECSNumber MO7875000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.