Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vinyl acetate, 99%, stab. with 8-12ppm hydroquinone

Catalog No. aaa16247ae
Click to view available options
Quantity:
100 mL
500 mL
2500 mL

CAS: 108-05-4 | C4H6O2 | 86.09 g/mol

Vinyl acetate is used in the preparation of polyvinyl acetate emulsions and polyvinyl alcohol. It acts as a precursor to polyvinyl acetate. It serves as an ingredient in furniture-glue. It is also used to prepare copolymers with other monomers such as ethylene, acrylic acid. Further, it is used to make adhesives, paints and plastics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Vinyl acetate is used in the preparation of polyvinyl acetate emulsions and polyvinyl alcohol. It acts as a precursor to polyvinyl acetate. It serves as an ingredient in furniture-glue. It is also used to prepare copolymers with other monomers such as ethylene, acrylic acid. Further, it is used to make adhesives, paints and plastics.

Solubility
Slightly miscible with water.

Notes
Light sensitive. Incompatible with acids, bases, oxidizing agents, peroxides, chlorosulfonic acid, ethylene imine, hydrochloric acid, oleum, nitric acid, sulfuric acid and 2-aminoethanol.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-05-4
Molecular Formula C4H6O2
Molecular Weight (g/mol) 86.09
MDL Number MFCD00008713
InChI Key XTXRWKRVRITETP-UHFFFAOYSA-N
Synonym vinyl acetate, acetic acid ethenyl ester, acetic acid vinyl ester, ethenyl ethanoate, 1-acetoxyethylene, vinyl ethanoate, acetoxyethene, vinylacetat, vinyl acetate monomer, vinyl a monomer
PubChem CID 7904
ChEBI CHEBI:46916
IUPAC Name ethenyl acetate
SMILES CC(=O)OC=C

Specifications

Melting Point -100°C
Density 0.932
Boiling Point 72°C to 73°C
Flash Point −8°C (18°F)
Odor Sweet
Refractive Index 1.396
Linear Formula CH2=CHO2CCH3
Quantity 100 mL
UN Number UN1301
Beilstein 1209327
Merck Index 14,9992
Solubility Information Slightly miscible with water.
Formula Weight 86.09
Percent Purity 99%
Chemical Name or Material Vinyl acetate, stabilized with 8 to 12ppm hydroquinone
Show More Show Less
Hazard Category H225-H332-H335-H351
Hazard Statement GHS H Statement
H225
Highly flammable liquid and vapour.
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P261-P271-P281-P303+P361+P353-P304+P340-P308+P313-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: VINYL ACETATE, STABILIZED
EINECSNumber 203-545-4
RTECSNumber AK0875000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.