Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vinyl decanoate, 95%, stab.

Catalog No. p-7030483
Click to view available options
Quantity:
5 g
25 g

CAS: 4704-31-8 | C12H22O2 | 198.306 g/mol

It is employed as a intermediates for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a intermediates for pharmaceutical.

Solubility
Soluble in methanol. Not miscible or difficult to mix with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4704-31-8
Molecular Formula C12H22O2
Molecular Weight (g/mol) 198.306
MDL Number MFCD00027349
InChI Key CMDXMIHZUJPRHG-UHFFFAOYSA-N
Synonym vinyl decanoate, decanoic acid, vinyl ester, vinyl n-decanoate, unii-ff00i399li, decanoic acid, ethenyl ester, n-capric acid vinyl ester, decanoic acid vinyl, acmc-1aqz4, decanoic acid vinyl ester, capric acid vinyl ester = vinyl decenoate
PubChem CID 62140
IUPAC Name ethenyl decanoate
SMILES CCCCCCCCCC(=O)OC=C

Specifications

Density 0.886
Boiling Point 119°C to 120°C (10 mmHg)
Flash Point 104°C (219°F)
Refractive Index 1.435
Linear Formula CH3(CH2)8CO2CH=CH2
Quantity 5 g
Solubility Information Soluble in methanol. Not miscible or difficult to mix with water.
Formula Weight 198.3
Percent Purity 95%
Chemical Name or Material Vinyl decanoate, stabilized
EINECSNumber 225-185-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.