Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Vinyl Duct Tape

Catalog No. p-7182389 Shop All 3M Products
Click to view available options
Color:
Blue
Gray
Green
Yellow

Catalog No. 19-072-109

Supplier: 3M™ 7100146009

Only null left
Add to Cart
Add to Cart

3M™ Vinyl Duct Tape is a hand-tearable general purpose tape with a rubber adhesive and embossed vinyl backing.

  • Rubber adhesive sticks fast and stays fast on many surfaces, even irregular surfaces
  • Embossed vinyl backing seals out dust, dirt, and moisture, and resists abrasion and many chemicals
  • Ideal for a variety of purposes including temporary repairs, identification, contrast, decoration, securing wiring, bundling, sealing, holding, protecting, and other jobs including applications requiring moisture resistance
  • Peels off cleanly after 30 minutes on painted steel as hot as 200°F (93°C)
  • Easy to tear and conforms to irregular surfaces
  • Available in a variety of colors for coding, process tagging, highlighting trip hazards, and enhancing décor
TRUSTED_SUSTAINABILITY

Specifications

Color Yellow
Product Type Vinyl Duct Tape
For Use With (Application) Communications, Design and Construction, Manufacturing, Mining, Oil and Gas, Transportation
Model 3903
Quantity 24/Cs.
Thickness (English) 6.5 mil
Dimensions (LxW) 50 yd. x 2 in.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.