Learn More
Vinylcyclohexane, 97%
Description
The use of vinylcyclohexane dioxide (VCD) as a polar dehydrant with subsequent embedment in Spurr was studied. Vinylcyclohexane (VCH) was copolymerized with ethene and propene using methylaluminoxane-activated metallocene catalysts.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
The use of vinylcyclohexane dioxide (VCD) as a polar dehydrant with subsequent embedment in Spurr was studied. Vinylcyclohexane (VCH) was copolymerized with ethene and propene using methylaluminoxane-activated metallocene catalysts.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Specifications
| Density | 0.805 |
| Boiling Point | 127°C to 128°C |
| Flash Point | 22°C (71°F) |
| Refractive Index | 1.4465 |
| Quantity | 5 g |
| UN Number | UN3295 |
| Beilstein | 1304663 |
| Solubility Information | Insoluble in water. |
| Formula Weight | 110.2 |
| Percent Purity | 97% |
| Show More |
Safety and Handling
| Hazard Category | H225 |
| Hazard Statement | GHS H Statement H225-H304 Highly flammable liquid and vapor. May be fatal if swallowed and enters airways. |
| Precautionary Statement | P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S. |
| EINECSNumber | 211-779-3 |
| RTECSNumber | GV7800000 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.