Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vinylcyclopentane, 97%

Catalog No. ac150050010
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle

CAS: 3742-34-5 | C7H12 | 96.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3742-34-5
Molecular Formula C7H12
Molecular Weight (g/mol) 96.17
MDL Number MFCD00001386
InChI Key BEFDCLMNVWHSGT-UHFFFAOYSA-N
Synonym vinylcyclopentane, cyclopentane, ethenyl, cyclopentane, vinyl, cyclopentyl-ethene, cyclopentylethylene, cyclopentane,ethenyl, acmc-209iss
PubChem CID 77345
IUPAC Name ethenylcyclopentane
SMILES C=CC1CCCC1

Specifications

Color Colorless
Density 0.7000g/mL
Boiling Point 97.0°C
Flash Point 10°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.436
Linear Formula C5H9CH=CH2
Quantity 1 g
Specific Gravity 0.7
Formula Weight 96.17
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Vinylcyclopentane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapour.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Ground/Bond container and receiving equipment.
EINECSNumber 223-133-8

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.