Learn More
Vinylene carbonate, 97+%, stab. with BHT
Description
Vinylene carbonate is used as an additive to electrolyte solutions for anode side Lithium-ion batteries. It also acts as a sealant to seal at least a portion of the silicon-polyvinyl acid interface. It is further used for great improvement of high temperature performance of the battery.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 19°C to 22°C |
| Color | Colorless to Yellow |
| Density | 1.355 g/mL |
| Boiling Point | 162°C to 163°C |
| Flash Point | 72°C (163°F) |
| Refractive Index | 1.421 |
| Quantity | 2 g |
| Sensitivity | Moisture sensitive |
| Solubility Information | Soluble in water. |
| Formula Weight | 86.05 |
| Show More |
Safety and Handling
| Hazard Category | H227-H302-H311-H315-H317-H318-H373 |
| Hazard Statement | GHS H Statement H302-H332-H315-H319-H317-H335-H227 Harmful if swallowed. Harmful if inhaled. Causes skin irritation. Causes serious eye irritation. May cause an allergic skin reaction. May cause respiratory irritation. Combustible liquid. |
| Precautionary Statement | P210-P260-P264b-P270-P272-P280-P301+P312-P302+P352-P305+P351+P338-P310-P312-P314-P330-P333+P313-P361-P363-P370+P378q-P501c |
| DOTInformation | Hazard Class: 6.1; Packaging Group: III |
| EINECSNumber | 212-825-5 |
| RTECSNumber | FG3325000 |
| TSCA | Yes |
| storageNote1 | 2°C to 8°C |
| Recommended Storage | Keep cold; Store under Argon |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.