Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vinylphosphonic Acid 95.0+%, TCI America™
SDP

Catalog No. V0068250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V0068250G 250 g
V006825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. V0068250G Supplier TCI America Supplier No. V0068250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1746-03-8
Molecular Formula C2H3O3P
Molecular Weight (g/mol) 106.02
MDL Number MFCD00043866
InChI Key ZTWTYVWXUKTLCP-UHFFFAOYSA-L
Synonym vinylphosphonic acid, phosphonic acid, ethenyl-, homopolymer, vinyl-phosphonicaci, ethenephosphonicacid, ethenylphosphonicacid, ethenyl-phosphonicaci, ethylenephosphonicacid, phosphonic acid, p-ethenyl-, homopolymer, acmc-1bvr6, poly vinylphosphonic acid
PubChem CID 168725
IUPAC Name ethenylphosphonate
SMILES [O-]P([O-])(=O)C=C

Specifications

Color Yellow
Boiling Point 98°C
Quantity 250 g
UN Number 3265
Formula Weight 108.03
Percent Purity ≥95.0% (T)
Physical Form Liquid
Chemical Name or Material Vinylphosphonic Acid
RTECSNumber SZ7903500
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.