Learn More
Vinyltriethoxysilane, 97%
Description
Vinyltriethoxysilane is used as an adhesive for glass slides. It acts as a coupling agent on inorganic substances blended into polymers that function as adhesion promoters, crosslinkers, and surface modifiers. Further, it is used to prepare polymers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Vinyltriethoxysilane is used as an adhesive for glass slides. It acts as a coupling agent on inorganic substances blended into polymers that function as adhesion promoters, crosslinkers, and surface modifiers. Further, it is used to prepare polymers.
Solubility
Miscible with organic solvents.
Notes
Moisture sensitive. Incompatible with alkali, acid, water and strong oxidizing agents.

Specifications
Specifications
Density | 0.903 |
Boiling Point | 160°C to 161°C |
Flash Point | 34°C (93°F) |
Odor | Fruit-like |
Assay Percent Range | 0.97 |
Refractive Index | 1.399 |
Linear Formula | (CH3CH2O)3SiCH=CH2 |
Quantity | 1000 g |
UN Number | UN1993 |
Beilstein | 1767229 |
Show More |
Safety and Handling
Hazard Category | H226 |
Hazard Statement | GHS H Statement H226-H319-H335 Flammable liquid and vapour. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S. |
EINECSNumber | 201-081-7 |
RTECSNumber | VV6700000 |
TSCA | Yes |
storageNote1 | Moisture Sensitive |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.