Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vinyltrimethoxysilane, 98%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
Packaging:
Glass bottle
2768-02-7
C5H12O3Si
148.24
MFCD00008605
NKSJNEHGWDZZQF-UHFFFAOYSA-N
vinyltrimethoxysilane,trimethoxyvinylsilane,silane, ethenyltrimethoxy,trimethoxy vinyl silane,trimethoxysilyl ethene,vinyl trimethoxy silane,silane, trimethoxyvinyl,trimethoxysilyl ethylene,vtmo,vts-m
76004
ethenyl(trimethoxy)silane
CO[Si](C=C)(OC)OC
2768-02-7
C5H12O3Si
148.24
MFCD00008605
NKSJNEHGWDZZQF-UHFFFAOYSA-N
vinyltrimethoxysilane,trimethoxyvinylsilane,silane, ethenyltrimethoxy,trimethoxy vinyl silane,trimethoxysilyl ethene,vinyl trimethoxy silane,silane, trimethoxyvinyl,trimethoxysilyl ethylene,vtmo,vts-m
76004
ethenyl(trimethoxy)silane
CO[Si](C=C)(OC)OC
Viscosity 0.6 mm2/s (25°C)
Packaging Glass bottle
Flash Point 23°C
Density 0.9680g/mL
Refractive Index 1.3915 to 1.3935
Quantity 500g
Boiling Point 123.0°C
Specific Gravity 0.968
Assay Percent Range 97.5% min. (GC)
Linear Formula H2C=CHSi(OCH3)3
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in most common organic solvents
Formula Weight 148.24
Percent Purity 98%
Chemical Name or Material Vinyltrimethoxysilane
Show More Show Less

GHS H Statement
Flammable liquid and vapor.
Harmful if inhaled.
May cause damage to organs through prolonged or repeated exposure.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest

GHS Signal Word: Warning

EINECSNumber : 220-449-8