Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vinyltrimethoxysilane, 98%

Catalog No. ac216521000
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 2768-02-7 | C5H12O3Si | 148.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2768-02-7
Molecular Formula C5H12O3Si
Molecular Weight (g/mol) 148.24
MDL Number MFCD00008605
InChI Key NKSJNEHGWDZZQF-UHFFFAOYSA-N
Synonym vinyltrimethoxysilane, trimethoxyvinylsilane, silane, ethenyltrimethoxy, trimethoxy vinyl silane, trimethoxysilyl ethene, vinyl trimethoxy silane, silane, trimethoxyvinyl, trimethoxysilyl ethylene, vtmo, vts-m
PubChem CID 76004
IUPAC Name ethenyl(trimethoxy)silane
SMILES CO[Si](C=C)(OC)OC

Specifications

Density 0.9680g/mL
Boiling Point 123.0°C
Flash Point 23°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula H2C=CHSi(OCH3)3
Refractive Index 1.3915 to 1.3935
Quantity 100 g
Specific Gravity 0.968
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in most common organic solvents
Viscosity 0.6 mm2/s (25°C)
Formula Weight 148.24
Percent Purity 98%
Chemical Name or Material Vinyltrimethoxysilane
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Flammable liquid and vapor.
Harmful if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest
EINECSNumber 220-449-8

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.