Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vinyltriphenylphosphonium bromide, 97%

Catalog No. AAL0070814
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 5044-52-0 | C20H18BrP | 369.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5044-52-0
Molecular Formula C20H18BrP
Molecular Weight (g/mol) 369.24
MDL Number MFCD00011807
InChI Key VRAYVWUMBAJVGH-UHFFFAOYSA-M
Synonym vinyltriphenylphosphonium bromide, triphenyl vinyl phosphonium bromide, schweizer's reagent, ethenyltriphenylphosphonium bromide, ethenyl triphenyl phosphanium bromide, ethenyltriphenylphosphanium bromide, triphenyl-vinyl-phosphonium; bromide, pubchem16334, acmc-1aw2n, vinyltriphenylphosphine bromide
PubChem CID 2723893
IUPAC Name ethenyl(triphenyl)phosphanium;bromide
SMILES [Br-].C=C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 182°C to 186°C
Quantity 25 g
Beilstein 4168860
Sensitivity Hygroscopic; Light sensitive
Formula Weight 369.25
Percent Purity 97%
Chemical Name or Material Vinyltriphenylphosphonium bromide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: II
EINECSNumber 225-740-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.