Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Viomycin
GREENER_CHOICESDP

Catalog No. 378710 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37-871-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 37-871-0 Supplier R&D Systems™ Supplier No. 3787/10

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Antibiotic; inhibits bacterial DNA synthesis

Viomycin is a member of the tuberactinomycin family of antibiotics. Inhibits group I intron splicing and prokaryotic protein synthesis. Freezes bacterial ribosomes in either the pre- or post-translational state. Facilitates trans-cleavage of the Neurospora VS ribozyme.

Chemical Identifiers

CAS 32988-50-4
Molecular Formula C25H43N13O10

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 881.85
Product Type Viomycin
Purity >98%
Molecular Weight (g/mol) 685.7
Chemical Name or Material Viomycin
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.