Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VIP Antibody (2) - BSA Free, Novus Biologicals™
SDP

Catalog No. p-7200543 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018334 0.025 mL
NBP105163 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018334 Supplier Novus Biologicals Supplier No. NBP1051630.025ML
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

VIP Monoclonal specifically detects VIP in Human, Rat, Porcine, Bovine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen VIP
Applications Immunohistochemistry (Paraffin), Immunohistochemistry, ELISA, Western Blot, Immunofluorescence, Immunocytochemistry
Classification Monoclonal
Clone 2
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, ELISA 1:1000, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:100
Formulation 10mM Phosphate (pH 7.4) and 0.5M NaCl with 0.05% Sodium Azide
Gene Accession No. P01282
Gene Alias MGC13587, PHM27, vasoactive intestinal peptide, VIP peptides
Gene Symbols VIP
Host Species Mouse
Immunogen Vasoactive intestinal peptide adsorbed onto aluminum hydroxide gel
Purification Method Protein A or G purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, GPCR, Neuroscience, Neurotransmission, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 7432
Test Specificity Binds to human, bovine, porcine or rat VIP, but does not bind to PHM or PACAP
Target Species Bovine, Pig, Rat, Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.