Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VIP Antibody (OTI1E8), Alexa Fluor™ 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274848AF488
Description
VIP Monoclonal specifically detects VIP in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
| VIP | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| VIP | |
| Full length recombinant protein of human VIP (NP_003372) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI1E8 | |
| Western Blot, Immunohistochemistry | |
| MGC13587, PHM27, vasoactive intestinal peptide, VIP peptides | |
| Mouse | |
| Affinity Purified | |
| Cancer, Cell Cycle and Replication, GPCR, Neuroscience, Neurotransmission, Signal Transduction | |
| 7432.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction