Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VIP Antibody (OTI1E8), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274848R
Description
VIP Monoclonal specifically detects VIP in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
VIP | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
VIP | |
Full length recombinant protein of human VIP (NP_003372) produced in E.coli. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI1E8 | |
Western Blot, Immunohistochemistry | |
MGC13587, PHM27, vasoactive intestinal peptide, VIP peptides | |
Mouse | |
Affinity Purified | |
Cancer, Cell Cycle and Replication, GPCR, Neuroscience, Neurotransmission, Signal Transduction | |
7432.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction