Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Scigen VIP-Fixative™ Non-precipitating Buffered Zinc 10% Formalin

Catalog No. p-4882129 Shop All Scigen Inc Products
Change view
Click to view available options
Capacity:
3.8 L
19 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity
23-730-584 19 L
23-730-583 3.8 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-730-584 Supplier Scigen Supplier No. 5043
This item is not returnable. View return policy

Compatible with and replaces traditional 10% NBF. Eliminates routine warm water flushes, and salt accumulation in all tissue processors. Neutralex™ compatible for Down-the-Drain disposal.

  • Introduced in 2001, VIP-Fixative and has been marketed under several well-known brands worldwide for use in automatic tissue processors as a non-precipitating formalin fixative
  • VIP-Fixative formula will not form solids (precipitate) in tissue processors and can eliminate the need for routine warm water flushing by preventing precipitate damage to the rotary valve.
  • Clear, colorless solution free of particles, buffered zinc 10% formalin based, for use in histological applications of fixation and storage of tissue specimens, soluble is dehydrant reagents
  • Improves cellular details and improves cell wall rigidity resulting in easier cutting and uniform sections.
  • Highly visible OSHA compliant labeling
  • Buffered zinc formalin enhances nuclear detail in immunohistochemistry and special staining techniques
  • Chloride, sulfate and mercury free
  • Nonsterile containers marked with a lot code and expiration date, 4 year shelf life
  • Formalin waste designed to be treated in bulk using Neutralex
  • Manufactured using ISO 13485 and FDA quality systems

Specifications

Capacity 19 L
Closure Type Lined Cap with Quick Release Faucet
Color Clear, Water White
Compatibility Compatible with Aqueous and Alcoholic Solutions
Content And Storage Room temperature
Concentration Buffered Zinc 10% Formalin
For Use With (Equipment) All Tissue Processors
Product Type Formalin
Solution Type Formalin
For Use With (Application) Tissue processor, to prevent instrument damage from solids with no impact on specimen integrity
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.