Learn More
VIP Mouse anti-Human, Clone: OTI1E8, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF806850

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The protein encoded by this gene belongs to the glucagon family. It stimulates myocardial contractility, causes vasodilation, increases glycogenolysis, lowers arterial blood pressure and relaxes the smooth muscle of trachea, stomach and gall bladder. Alternative splicing occurs at this locus and two transcript variants encoding distinct isoforms have been identified.Specifications
VIP | |
Monoclonal | |
Unconjugated | |
VIP | |
PHM27 | |
Mouse | |
Affinity Chromatography | |
RUO | |
7432 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI1E8 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P01282 | |
VIP | |
Full length human recombit protein of human VIP produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.