Learn More
Invitrogen™ VIP Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA594931
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - IHC: human pancreatic cancer tissue, rat pancreas tissue, rat intestine tissue, mouse pancreas tissue, mouse intestine tissue. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
VIP encodes a protein that belongs to the glucagon family. It stimulates myocardial contractility, causes vasodilation, increases glycogenolysis, lowers arterial blood pressure and relaxes the smooth muscle of trachea, stomach and gall bladder. Alternative splicing occurs at this locus and two transcript variants encoding distinct isoforms have been identified.
Specifications
VIP | |
Polyclonal | |
Unconjugated | |
VIP | |
Intestinal peptide PHI-27; Intestinal peptide PHI-42; Intestinal peptide PHM-27; Intestinal peptide PHV-42; MGC13587; PACAP-R-2; Peptide histidine isoleucinamide 27; Peptide histidine methioninamide 27; Peptide histidine valine 42; PHI; PHI, peptide histidine isoleucine; PHM27; prepro-VIP; vasoactive intestinal peptide; Vasoactive intestinal polypeptide; VIP; VIP peptides; vip/phi27; VIP-R-1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
117064, 22353, 7432 | |
-20°C | |
Lyophilized |
Immunohistochemistry (Paraffin) | |
500 μg/mL | |
PBS with 5mg BSA and 0.05mg sodium azide | |
P01282, P01283, P32648 | |
VIP | |
A synthetic peptide corresponding to a sequence in the middle region of human VIP (81-107aa HADGVFTSDFSKLLGQLSAKKYLESLM). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.