Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VIP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111702
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111702 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111702 Supplier Invitrogen™ Supplier No. PA5111702
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AWPLYRAPSA LRLGDRIPFE Highest antigen sequence indentity to the following orthologs: Rat - 65%, Mouse - 62%.

VIP encodes a protein that belongs to the glucagon family. It stimulates myocardial contractility, causes vasodilation, increases glycogenolysis, lowers arterial blood pressure and relaxes the smooth muscle of trachea, stomach and gall bladder. Alternative splicing occurs at this locus and two transcript variants encoding distinct isoforms have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VIP
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene VIP
Gene Accession No. P01282
Gene Alias Intestinal peptide PHI-27; Intestinal peptide PHI-42; Intestinal peptide PHM-27; Intestinal peptide PHV-42; MGC13587; PACAP-R-2; Peptide histidine isoleucinamide 27; Peptide histidine methioninamide 27; Peptide histidine valine 42; PHI; PHI, peptide histidine isoleucine; PHM27; prepro-VIP; vasoactive intestinal peptide; Vasoactive intestinal polypeptide; VIP; VIP peptides; vip/phi27; VIP-R-1
Gene Symbols VIP
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human VIP. Recombinant protein control fragment (Product #RP-107894).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7432
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.