Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VIP Receptor 1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501985060
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-5060 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-5060 Supplier Bioss Supplier No. BS2982R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The Vasoactive Intestinal Polypeptide Receptor 1 (VPAC1) is a member of a family of three G-protein coupled receptors which mediate VIP (vasoactive intestinal peptide) and PACAP (pituitary adenlyate cyclase activating peptide) function in a variety of tissues. Together, the VPAC1 and VPAC2 receptors compose a subtype of the VIP receptor. Expression of the VIP and PACAP receptors has been linked to the presence of human tumors.

Specifications

Antigen VIP Receptor 1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene VIPR1
Gene Accession No. P32241
Gene Alias AV071699; HVR1; II; Pacap receptor; PACAP type II receptor; PACAP-R2; PACAP-R-2; pituitary adenylate cyclase activating polypeptide receptor, type II; Pituitary adenylate cyclase-activating polypeptide type II receptor; Pvr2; RDC1; type 1 vasoactive intestinal peptide receptor; V1RG; VAPC1; vasoactive intestinal peptide receptor 1; Vasoactive intestinal polypeptide receptor 1; VIP and PACAP receptor 1; VIP receptor 1; VIP receptor subtype 1; VIP receptor, type I; VIP1 receptor; VIPR; Vipr1; VIP-R1; VIP-R-1; VIRG; VPAC1; VPAC1 receptor; VPAC1R; VPCAP1R
Gene Symbols VIPR1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human VIP Receptor 1/VAPC1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7433
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.