Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ VIP36 Monoclonal Antibody (02)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA529395
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human LMAN2/VIP36.
This gene encodes a type I transmembrane lectin that shuttles between the endoplasmic reticulum, the Golgi apparatus and the plasma membrane. The encoded protein binds high mannose type glycoproteins and may facilitate their sorting, trafficking and quality control.
Specifications
VIP36 | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
Q12907 | |
LMAN2 | |
Recombinant Human LMAN2/VIP36 protein (Asp45-Arg322). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
ELISA, Immunoprecipitation, Western Blot | |
02 | |
Unconjugated | |
LMAN2 | |
1110003H06Rik; 1300009F09Rik; AA408240; AL023023; AU040819; C5orf8; glycoprotein GP36b; GP36B; Lectin mannose-binding 2; lectin, mannose binding 2; lectin, mannose-binding 2; Lman2; vesicular integral protein of 36 kDa; Vesicular integral-membrane protein 36; Vesicular integral-membrane protein VIP36; Vip36; VIP36 (vesicular integral-membrane protein); wu:fb16f04; zgc:158761 | |
Mouse | |
Protein A | |
RUO | |
10960 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction