Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VIP36 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5119251
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5119251 200 μL
PIPA5121508 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5119251 Supplier Invitrogen™ Supplier No. PA5119251
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

VIP36 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

This gene encodes a type I transmembrane lectin that shuttles between the endoplasmic reticulum, the Golgi apparatus and the plasma membrane. The encoded protein binds high mannose type glycoproteins and may facilitate their sorting, trafficking and quality control.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VIP36
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene LMAN2
Gene Accession No. Q12907
Gene Alias 1110003H06Rik; 1300009F09Rik; AA408240; AL023023; AU040819; C5orf8; glycoprotein GP36b; GP36B; Lectin mannose-binding 2; lectin, mannose binding 2; lectin, mannose-binding 2; Lman2; vesicular integral protein of 36 kDa; Vesicular integral-membrane protein 36; Vesicular integral-membrane protein VIP36; Vip36; VIP36 (vesicular integral-membrane protein); wu:fb16f04; zgc:158761
Gene Symbols LMAN2
Host Species Rabbit
Immunogen Recombinant protein Lectin, Mannose Binding 2. The antigen corresponds to amino acid range 45-322 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10960, 290994
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.