Learn More
Invitrogen™ VIP36 Polyclonal Antibody

Description
Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%.
Specifications
Specifications
| Antigen | VIP36 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 |
| Gene | LMAN2 |
| Gene Accession No. | Q12907 |
| Gene Alias | 1110003H06Rik; 1300009F09Rik; AA408240; AL023023; AU040819; C5orf8; glycoprotein GP36b; GP36B; Lectin mannose-binding 2; lectin, mannose binding 2; lectin, mannose-binding 2; Lman2; vesicular integral protein of 36 kDa; Vesicular integral-membrane protein 36; Vesicular integral-membrane protein VIP36; Vip36; VIP36 (vesicular integral-membrane protein); wu:fb16f04; zgc:158761 |
| Gene Symbols | LMAN2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.