Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VIPAR Antibody, Janelia Fluor™ 646, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP204091JF646
Description
VIPAR Polyclonal antibody specifically detects VIPAR in Human, Mouse samples. It is validated for Western BlotSpecifications
VIPAR | |
Polyclonal | |
Western Blot | |
C14orf133FLJ12707, hSPE-39, Protein spe-39 homolog, SPE-39, SPE39chromosome 14 open reading frame 133, VPS16B, VPS33B interacting protein, apical-basolateral polarity regulator, VPS33B-interacting protein, VPS33B-interacting protein involved in polarity and apical protein restriction | |
The immunogen recognized by this antibody maps to a region between residue 443 and 493 of human VPS33B Interacting Protein, Apical-Basolateral Polarity Regulator using the numbering given in entry NP_071350.2 (GeneID 63894). | |
0.1 mL | |
Primary | |
Human, Mouse | |
Purified |
Western Blot | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
63894 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction