Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VIPAR Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501565379
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-5379 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-5379 Supplier Bethyl Laboratories, Inc Supplier No. A303526AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

Proposed to be involved in endosomal maturation implicating in part VPS33B. In epithelial cells, the VPS33B:VIPAS39 complex may play a role in the apical RAB11A-dependent recycling pathway and in the maintece of the apical-basolateral polarity. May play a role in lysosomal trafficking, probably via association with the core HOPS complex in a discrete population of endosomes; the functions seems to be independent of VPS33B. May play a role in vesicular trafficking during spermatogenesis. May be involved in direct or indirect transcriptional regulation of E-cadherin.

Specifications

Antigen VIPAR
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene VIPAS39
Gene Accession No. Q8BGQ1, Q9H9C1
Gene Alias C14orf133, chromosome 14 open reading frame 133, hSPE-39, SPE39, SPE-39, VIPAR, VPS16B, VPS33B interacting protein, apical-basolateral polarity regulator, VPS33B-interacting protein in apical-basolateral polarity regulator, VPS33B-interacting protein in polarity and apical restriction, VPS33B-interacting protein involved in polarity and apical protein restriction
Gene Symbols VIPAS39
Host Species Rabbit
Immunogen Between 443 and 493
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 104799, 63894
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.