Learn More
VIPAR Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303526A

Description
The recommended shelf life for this product is 1 year from date of receipt.
Proposed to be involved in endosomal maturation implicating in part VPS33B. In epithelial cells, the VPS33B:VIPAS39 complex may play a role in the apical RAB11A-dependent recycling pathway and in the maintece of the apical-basolateral polarity. May play a role in lysosomal trafficking, probably via association with the core HOPS complex in a discrete population of endosomes; the functions seems to be independent of VPS33B. May play a role in vesicular trafficking during spermatogenesis. May be involved in direct or indirect transcriptional regulation of E-cadherin.Specifications
VIPAR | |
Polyclonal | |
Unconjugated | |
VIPAS39 | |
C14orf133, chromosome 14 open reading frame 133, hSPE-39, SPE39, SPE-39, VIPAR, VPS16B, VPS33B interacting protein, apical-basolateral polarity regulator, VPS33B-interacting protein in apical-basolateral polarity regulator, VPS33B-interacting protein in polarity and apical restriction, VPS33B-interacting protein involved in polarity and apical protein restriction | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
104799, 63894 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q8BGQ1, Q9H9C1 | |
VIPAS39 | |
Between 443 and 493 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.