Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VIPAS39 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551971
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551971 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551971 Supplier Invitrogen™ Supplier No. PA551971
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AHIQDHYTLL ERQIIIEAND RHLESAGQTE IFRKHPRKAS ILNMPLVTTL FYSCFYHYTE AEGTFSSPVN LKKTFKIPDK QYVLTALAAR AKLRAWNDVD ALFTTKNWLG YTKKRAPIGF HRVVEILHKN NAPVQILQEY VNLVEDVDTK Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.

This gene encodes a protein involved in the sorting of lysosomal proteins. Mutations in this gene are associated with ARCS2 (arthrogryposis, renal dysfunction, and cholestasis-2). Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VIPAS39
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene VIPAS39
Gene Accession No. Q9H9C1
Gene Alias 6720456H09Rik; 9330175H22Rik; AI413782; C10H14orf133; C14orf133; C8H14orf133; hSPE-39; protein spe-39 homolog; SPE39; SPE-39; SPE-39 protein; Spermatogenesis-defective protein 39 homolog; VIPAR; VIPAS39; VPS16B; VPS33B interacting protein, apical-basolateral polarity regulator, spe-39 homolog; VPS33B-interacting protein in apical-basolateral polarity regulator; VPS33B-interacting protein in polarity and apical restriction; VPS33B-interacting protein involved in polarity and apical protein restriction
Gene Symbols VIPAS39
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human VIPAS39. Recombinant protein control fragment (Product #RP-89056).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 63894
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.