Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Viperin Mouse anti-Human, Mouse, Clone: MaP.VIP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP307918
Description
Viperin Monoclonal antibody specifically detects Viperin in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation, ImmunofluorescenceSpecifications
Viperin | |
Monoclonal | |
0.1 mg/mL | |
PBS containing 0.1% BSA | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Western Blot, Flow Cytometry, Immunoprecipitation, Immunofluorescence | |
MaP.VIP | |
Unconjugated | |
2510004L01Rik, cig33, cig5, Cytomegalovirus-induced gene 5 protein, interferon-inducible, radical S-adenosyl methionine domain containing 2, radical S-adenosyl methionine domain-containing protein 2, vig1, Viperin, Virus inhibitory protein, endoplasmic reticulum-associated | |
Recombinant mouse viperin fragment (residues 92-362) | |
50 μg | |
Immunology | |
91543 | |
Store at 4°C. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction