Learn More
Invitrogen™ Viperin Polyclonal Antibody

Description
Immunogen sequence: YLDILAISCDS FDEEVNVLIG RGQGKKNHVE NLQKLRRWCR DYRVAFKINS VINRFNVEED MTEQIKALNP VRWKVFQCLL IEGENCGEDA L Highest antigen sequence identity to the following orthologs - mouse 91%, rat 91%.
Specifications
Specifications
| Antigen | Viperin |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | Rsad2 |
| Gene Accession No. | Q8WXG1 |
| Gene Alias | 2510004L01Rik; Best5; Bone-expressed sequence tag 5 protein; cig33; cig5; CIG6; cytomegalovirus-induced gene 5 protein; hypothetical protein LOC570456; inflammatory response gene 6 protein; inflammatory response protein 6; IRG6; radical S-adenosyl methionine domain containing 2; radical S-adenosyl methionine domain-containing protein 2; rsad2; rsad2 {ECO:0000250; si:ch211-276e8.2; similar to radical S-adenosyl methionine domain containing 2; UniProtKB:Q8WXG1}; VHSV-induced-like protein; Vig1; Viperin; viral hemorrhagic septicemia virus (VHSV)-induced gene 1; virus inhibitory protein, endoplasmic reticulum-associated, interferon-inducible; zgc:112342 |
| Gene Symbols | Rsad2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.