Learn More
Invitrogen™ Viperin Polyclonal Antibody

Description
The amino acid sequence used as immunogen is 100% homologous in white-cheeked gibbon, chinese hamster, common chimpanzee, new world monkey, panda, african bush elephant and orangutan, 95% in horse, wild turkey, opossum, platypus, 89% in western clawed frog, rabbit, zebra finch, chicken, 79% in carolina anole, and 63% homologous in zebrafish and sablefish.
Specifications
Specifications
| Antigen | Viperin |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA and 0.05% sodium azide |
| Gene | Rsad2 |
| Gene Accession No. | O70600, Q2HJF9, Q8CBB9, Q8WXG1, Q9MZU4 |
| Gene Alias | 2510004L01Rik; Best5; Bone-expressed sequence tag 5 protein; cig33; cig5; CIG6; cytomegalovirus-induced gene 5 protein; hypothetical protein LOC570456; inflammatory response gene 6 protein; inflammatory response protein 6; IRG6; radical S-adenosyl methionine domain containing 2; radical S-adenosyl methionine domain-containing protein 2; rsad2; rsad2 {ECO:0000250; si:ch211-276e8.2; similar to radical S-adenosyl methionine domain containing 2; UniProtKB:Q8WXG1}; VHSV-induced-like protein; Vig1; Viperin; viral hemorrhagic septicemia virus (VHSV)-induced gene 1; virus inhibitory protein, endoplasmic reticulum-associated, interferon-inducible; zgc:112342 |
| Gene Symbols | Rsad2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.