Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Viral Chikungunya Virus E1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $514.50
Specifications
Antigen | ChikungunyaVirusE1 |
---|---|
Clone | 988122 |
Dilution | Western Blot 2μg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB97792SP
![]() |
R&D Systems
MAB97792SP |
25 μg |
Each for $200.50
|
|
|||||
MAB97792100
![]() |
R&D Systems
MAB97792100 |
100 μg |
Each for $514.50
|
|
|||||
Description
Chikungunya Virus E1 Monoclonal specifically detects Chikungunya Virus E1 in Viral samples. It is validated for Western Blot.Specifications
ChikungunyaVirusE1 | |
Western Blot 2μg/mL | |
Monoclonal | |
Supernatant | |
Virus | |
Q8JUX5 | |
Human embryonic kidney cell line HEK293-derived transfected with viral Chikungunya Virus E1 Tyr810-Gly1224 Accession # Q8JUX5 | |
Primary | |
Reconstitute at 0.5mg/mL in sterile PBS. | |
Detects viral Chikungunya Virus E1 in ELISAs. |
988122 | |
Western Blot | |
Unconjugated | |
Mouse | |
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
CHICKV E1, Chikungunya Virus, CHIKV-E1 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title