Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Viral Chikungunya Virus E1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$188.28 - $544.12
Specifications
| Antigen | ChikungunyaVirusE1 |
|---|---|
| Clone | 988110 |
| Dilution | ELISA |
| Applications | ELISA |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB9779SP
![]() |
R&D Systems
MAB9779SP |
25 μg |
Each for $188.28
|
|
|||||
MAB9779100
![]() |
R&D Systems
MAB9779100 |
100 μg |
Each for $544.12
|
|
|||||
Description
Chikungunya Virus E1 Monoclonal specifically detects Chikungunya Virus E1 in Viral samples. It is validated for ELISA.Specifications
| ChikungunyaVirusE1 | |
| ELISA | |
| Monoclonal | |
| Supernatant | |
| Virus | |
| Q8JUX5 | |
| Human embryonic kidney cell line HEK293-derived transfected with viral Chikungunya Virus E1 Tyr810-Gly1224 Accession # Q8JUX5 | |
| Primary | |
| Reconstitute at 0.5mg/mL in sterile PBS. | |
| Detects viral Chikungunya Virus E1 in direct ELISAs. |
| 988110 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
| CHICKV E1, Chikungunya Virus, CHIKV-E1 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title