Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Viral CMV UL146/vCXC1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB620SP
This item is not returnable.
View return policy
Description
UL146/vCXC1 Monoclonal specifically detects UL146/vCXC1 in Viral samples. It is validated for Western Blot, Neutralization.Specifications
| UL146/vCXC1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| E. coli-derived recombinant viral CMV UL146/vCXC1 | |
| 25 μg | |
| Primary | |
| Detects viral CMV UL146/vCXC1 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human CXCL1, 2, 3, 5, 6, 7, 8, 9, 10, 11, 12, 13, recombinant mouse CXCL1, 2, 6, 9, 10, 12, recombinant rat CXCL1, 3, recombinant viral MIP-I, -II, or -III is observed. | |
| Virus | |
| Purified |
| Western Blot, Neutralization | |
| 76520 | |
| Western Blot 1 ug/mL, Neutralization 1.5-7.5 ug/mL | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 3077566 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction