Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Viral CMV UL147/vCXC2 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF606
This item is not returnable.
View return policy
Description
UL147/vCXC2 Polyclonal specifically detects UL147/vCXC2 in Viral samples. It is validated for Western Blot.Specifications
UL147/vCXC2 | |
Polyclonal | |
Unconjugated | |
UL147 | |
E. coli-derived recombinant viral CMV UL147/vCXC2 | |
100 μg | |
Primary | |
Detects viral CMV UL147/vCXC2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant mouse (rm) Teck, recombinant human MIG, recombinant rat MIP-3a, and rmMIP-3b is observed. | |
Virus | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
Goat | |
Affinity Purified | |
RUO | |
3077419 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction