Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Viral Dengue Virus 1 NS1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $410.00
Specifications
Antigen | Dengue virus NS1 protein |
---|---|
Clone | 973602 |
Dilution | ELISA |
Applications | ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB9438SP
![]() |
R&D Systems
MAB9438SP |
25 μg |
Each for $200.50
|
|
|||||
MAB9438100
![]() |
R&D Systems
MAB9438100 |
100 μg |
Each for $410.00
|
|
|||||
Description
Dengue Virus NS1 protein Monoclonal specifically detects Dengue Virus NS1 protein in Viral samples. It is validated for ELISA.Specifications
Dengue virus NS1 protein | |
ELISA | |
Monoclonal | |
Supernatant | |
Virus | |
P17763 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
973602 | |
ELISA | |
Unconjugated | |
Mouse | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
Human embryonic kidney cell line HEK293-derived viral Dengue Virus 1 NS1 protein, Asp776-Ala1127, Accession No. P17763 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects viral Dengue Virus 1 NS1 protein in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title