Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Viral Dengue Virus NS1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $514.50
Specifications
Antigen | Dengue Virus NS1 |
---|---|
Clone | 971305 |
Dilution | Western Blot 2 ug/mL, ELISA |
Applications | Western Blot, ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB94441SP
![]() |
R&D Systems
MAB94441SP |
25 μg |
Each for $200.50
|
|
|||||
MAB94441100
![]() |
R&D Systems
MAB94441100 |
100 μg |
Each for $514.50
|
|
|||||
Description
Dengue Virus NS1 Monoclonal specifically detects Dengue Virus NS1 in Viral samples. It is validated for Western Blot, ELISA.Specifications
Dengue Virus NS1 | |
Western Blot 2 ug/mL, ELISA | |
Monoclonal | |
Supernatant | |
Virus | |
NP_740318 | |
Human embryonic kidney cell line HEK293-derived recombinant Dengue Virus 4 NS1 protein Asp1-Ala352 Accession # NP_740318 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects Dengue Virus 2/3/4 NS1 in Western blots. In sandwich immunoassays, Dengue Virus 1/2/3/4 NS1 is detected. |
971305 | |
Western Blot, ELISA | |
Unconjugated | |
Mouse | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
DEN-1, DEN-2, DEN-3, DEN-4, Dengue NS1, Dengue NS1 protein, Dengue Virus, Dengue virus Serotype 1, Dengue virus Serotype 2, Dengue virus Serotype 3, Dengue virus Serotype 4, Dengue virus Type 1, Dengue virus Type 2, Dengue virus Type 3, Dengue virus Type 4, DENV1, DENV1-NS1, DENV2, DENV2-NS1, DENV3, DENV3-NS1, DENV4, DENV4-NS1 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title