Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Viral EBV IL-10 Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $579.81
Specifications
| Antigen | IL-10 |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
IL-10 Polyclonal specifically detects IL-10 in Viral samples. It is validated for Western Blot.Specifications
| IL-10 | |
| Western Blot 0.1 ug/mL | |
| Polyclonal | |
| Goat | |
| Virus | |
| CSIF, CSIFMGC126450, Cytokine synthesis inhibitory factor, GVHDS, IL10, IL10A, IL-10MGC126451, interleukin 10, interleukin-10, TGIF | |
| CeHV15gBCRF1 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| P03180 | |
| 3586 | |
| E. coli-derived recombinant viral EBV IL-10 Gln26-Arg170 Accession # P03180 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects viral EBV IL-10 in direct ELISAs and Western blots. In direct ELISAs, approximately 35% cross-reactivity with recombinant canine IL-10 and recombinant human IL-10 is observed and less than 10% cross-reactivity with recombinant mouse IL-10 and recombinant rat IL-10 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title