Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Streck Viral Extraction Kit
SDP

Catalog No. 23285981
Click to view available options
:
Viral Extraction Kit, Format: Liquid
This item is not returnable. View return policy
This item is not returnable. View return policy

The Streck Viral Extraction Kit, powered by Ceres Nanotrap™ technology, is an efficient way to extract viral RNA from laboratory samples. Once isolated, the viral RNA can be used for qPCR, ddPCR and NGS analysis.

  • Ceres Nanotrap technology enhance viral extraction.
  • 96-sample configuration for automated protocol.
  • No additional chemicals required throughout workflow.
  • Ready-to-use buffers (no reconstitution necessary).
  • Increased sensitivity and specificity for viral identification.
  • Streamlined method to track new variants for epidemiological studies.

Specifications

Isolation Technology Magnetic Particles
Sample Type Wastewater
Sequencing Type RT-PCR and Digital PCR Sequencing
Product Type Viral Extraction Kit
Form Liquid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.