Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Viral HHV8-ORF74 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB5264
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5264 100 μg
MAB5264SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB5264 Supplier R&D Systems Supplier No. MAB5264
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

HHV8-ORF74 Monoclonal specifically detects HHV8-ORF74 in Viral samples. It is validated for Flow Cytometry, CyTOF-ready.

Specifications

Antigen HHV8-ORF74
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 462510
Concentration LYOPH
Conjugate Unconjugated
Dilution Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. AAB51506
Gene Symbols HHV8GK18_gp84
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with viral HHV8-ORF74 Met1-Thr342 Accession # AAB51506
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4961460
Test Specificity Detects viral HHV8-ORF74.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Virus
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.