Learn More
Viral HIV-1 Gag p24 Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | HIV-1 Gag p24 |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Clone | 1042960 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1 ug/mL, ELISA |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. |
| Gene Accession No. | Q77YG1_9HIV1 |
| Host Species | Mouse |
| Immunogen | E. coli-derived recombinant viral HIV-1 Gag p24, Pro149-Gly354, Accession # Q77YG1_9HIV1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.