Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Viral Japanese Encephalitis virus NS1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB10006SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB10006SP 25 μg
MB10006100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB10006SP Supplier R&D Systems Supplier No. MAB10006SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Japanese Encephalitis virus NS1 Monoclonal specifically detects Japanese Encephalitis virus NS1 in Viral samples. It is validated for Western Blot.

Specifications

Antigen Japanese Encephalitis virus NS1
Applications Western Blot
Classification Monoclonal
Clone 995010
Conjugate Unconjugated
Dilution Western Blot 1 μg;/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. P32886
Gene Alias Japanese Encephalitis, JE virus, JEV, JEVgp1, JEV-NS1
Host Species Mouse
Immunogen Human embryonic kidney cell, HEK293-derived Japanese Encephalitis virus NS1, Thr796-Ala1146, Accession No. P32886
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Primary or Secondary Primary
Gene ID (Entrez) 1489713
Test Specificity Detects Japanese Encephalitis virus NS1 in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Virus
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Lyophilized
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.