Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Viral Japanese Encephalitis virus NS1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $444.50
Specifications
Antigen | Japanese Encephalitis virus NS1 |
---|---|
Clone | 995026 |
Dilution | ELISA |
Applications | ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MB100062SP
![]() |
R&D Systems
MAB100062SP |
25 μg |
Each for $200.50
|
|
|||||
MB100062100
![]() |
R&D Systems
MAB100062100 |
100 μg |
Each for $444.50
|
|
|||||
Description
Japanese Encephalitis virus NS1 Monoclonal specifically detects Japanese Encephalitis virus NS1 in Viral samples. It is validated for ELISA.Specifications
Japanese Encephalitis virus NS1 | |
ELISA | |
Monoclonal | |
Mouse | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
Japanese Encephalitis, JE virus, JEV, JEVgp1, JEV-NS1 | |
Human embryonic kidney cell, HEK293-derived viral Japanese Encephalitis virus NS1 protein, Thr796-Ala1146, Accession No. P32886 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects viral Japanese Encephalitis virus NS1 protein in direct ELISAs. |
995026 | |
ELISA | |
Unconjugated | |
Virus | |
P32886 | |
1489713 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title