Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Viral MIP-II Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF601
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF601 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF601 Supplier R&D Systems Supplier No. BAF601
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

MIP-II Polyclonal specifically detects MIP-II in Viral samples. It is validated for Western Blot.

Specifications

Antigen MIP-II
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 1-2 ug/mL
Gene Accession No. NP_572064
Host Species Goat
Immunogen E. coli-derived recombinant human Viral MIP-II Leu24-Arg94 Accession # NP_572064
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Detects Viral MIP-II in Western blots. In Western blot, no cross-reactivity with recombinant human (rh) MIP-1 alpha , rhMIP-1 beta , recombinant viral (rv) MIP-I, and rvMIP-III is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Virus
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.